0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Standardy
Standardy organické
5-Hydroxyflunixin, analytical standard, VETRANAL™, Supelco®
5-Hydroxyflunixin, analytical standard, VETRANAL™, Supelco®
Catalog # 32463-10MG
Supplier:
Merck
CAS Number:
Expand image
5-Hydroxyflunixin, analytical standard, VETRANAL™, Supelco®
Catalog # 32463-10MG
Supplier:
Merck
CAS Number:
Specifications
Bal.:
10 mg
Standard:
analytical standard
Čí. CAS:
75369-61-8
Popis:
5-Hydroxyflunixin
Popis:
5-Hydroxyflunixin
Specifications
About this item
Product Details
Product Details