N-Vanillylnonanamide, analytical standard, Supelco®
Catalog # 15971-50MG
Supplier: Merck
CAS Number:
Specifications
- Bal.:50 mg
- Standard:Analytical standard
- Čí. CAS:2444-46-4
- Popis:N-Vanillylnonanamide ≥98% (HPLC)
- Vzorec:C₁₇H₂₇NO₃
- Skladovací teplota:2…8 °C
- Popis:N-Vanillylnonanamide