ContactUsLinkComponent
Colour Reference Solutions BY7 acc. to Ph, Supelco®
Colour Reference Solutions BY7 acc. to Ph, Supelco®
Katalogové číslo 71983-100ML-F
Supplier:  Merck
CAS Number:  
Colour Reference Solutions BY7 acc. to Ph, Supelco®
Katalogové číslo 71983-100ML-F
Supplier:  Merck
CAS Number:  
Specifications
  • Bal.:
    100 mL
  • Popis:
    Color Reference Solutions BY7