ContactUsLinkComponent
Gas generator, PG28L Purge
  PEAK45-1060EU
undefined
Gas generator, PG28L Purge
  PEAK45-1060EU
  • Průtoková rychlost:
    28 l/min
  • Popis:
    PG28L Purge Gas Generator, EU plug
  • Sekvence:
    01
  • Š×H×V:
    250×160×900 mm
  • Elektrické napětí:
    240 VAC, 50/60 Hz, 0,08 A
  • Tlak:
    6,89 bar (Outlet);
    6,89 to 8,27 bar (Inlet)
  • Hmotnost:
    45 kg
  • Bal.:
    1
The PG28L gas generator is suitable for laboratory detectors requiring CO₂ free gas such as FT-IR analysis. Using Pressure Swing Adsorption technology to remove organic contaminants and moisture from air, these generators provide a consistent flow of air to ensure little background noise in laboratory analysis.
  • Purge Gas (PG) generator offers carbon dioxide free gas
  • Best suited for commercial laboratories with a high throughput rate of samples and trials
  • PSA ensure maximum reduction of moisture (<–70 °C dew point)
  • Few moving parts means minimum maintenance requirements and 24/7 performance
  • Wall mountable for small laboratories
  • More cost effective than any other gas supply as the FT-IR detector needs a constant flow of gas
  • 1 year comprehensive on-site warranty
Purity: <1 ppm CO₂