ContactUsLinkComponent
Zero air generator, CG15L
Zero air generator, CG15L
Katalogové číslo PEAK43-1020
Dodavatel:  PEAK SCIENTIFIC
undefined
Zero air generator, CG15L
Katalogové číslo PEAK43-1020
Dodavatel:  PEAK SCIENTIFIC
Specifikace
  • Průtoková rychlost:
    15 l/min
  • Popis:
    Calibration Gas Generator, CG15L, EU plug
  • Sekvence:
    02
  • Š×H×V:
    410×430×410 mm
  • Bal.:
    1
Specifikace
O tomto produktu
CG15L generates zero air by removing carbon dioxide and moisture from the air for applications requiring a higher performance than zero air models.
  • Produces clean, dry laboratory grade air, ensures optimum supply for gas generators and instruments
  • Continuous supply of calibration gas
  • Completely silent operation
  • 1 year comprehensive on-site warranty
  • 1×1/4" BSP female gas outlet
  • Purity: <0,1 ppm THC
The gas produced by this range of generators exceeds performance levels of Zero Air and is ideal for a wide variety of air emission applications. Gas is generated by both catalytic oxidation and a proprietary regenerative sieve. Coalescing pre-filters and a final filter ensure continuous gas purity.
Dew point: –75° C
Inlet Pressure: 8,27 to 8,62 bar
Outlet Pressure: 5,52 to 6,89 bar
Power Consumption: 1100/540 W
Weight: 58 kg
Certifications: Meets requirements of Federal Register Specification 40CFR 86.114-79