Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, trans-Quercus lactone, analytical standard, Supelco®
Catalog # 72777-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, trans-Quercus lactone, analytical standard, Supelco®
Catalog # 72777-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Bal.:
    100 mg
  • Standard:
    Analytical standard
  • Čí. CAS:
    39638-67-0
  • Popis:
    trans-Quercus lactone, ≥95,0% (GC)
  • Vzorec:
    C₉H₁₆O₂
  • Popis:
    trans-Quercus lactone

Specifications

About this item