Supelco® Organic Reference Standard, Residual Solvents in Packaging Material Mixture 2
Catalog # 48995-U
Dodavatel: Merck
Číslo CAS:
Specifikace
- Koncentrace:9,09 % (v/v)
- Bal.:1 mL
- Standard:Certified reference material
- Popis:Residual solvents in packaging material mixture 2
- Podmínky uskladnění:2…8 °C
- Popis rizikové třídy:H225 - H318 - H336 - H351 - H360FD - H371
- Podmínky k odeslání:STD
- M-Clarity quality level:MQ100
- Bod vzplanutí:–18 °C
Specifikace
O tomto produktu
- GHS signal: GHS02, GHS08, GHS05, GHS07
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Caution: P202 - P210 - P233 - P280 - P305 + P351 + P338 - P308 + P311