Supelco® Organic Reference Standard, Residual Solvents in Packaging Material Mixture 1
Catalog # 48994-U
Supplier: Merck
CAS Number:
Specifications
- Koncentrace:7,14 % (v/v), pkg of 1 ml
- Bal.:1 mL
- Standard:Certified reference material
- Popis:Residual solvents in packaging material mixture 1
- Podmínky uskladnění:2…8 °C
- Popis rizikové třídy:H225 - H302 + H332 - H304 - H315 - H318 - H335 - H336 - H360FD - H371 - H411
- Podmínky k odeslání:STD
- M-Clarity quality level:MQ100
- Bod vzplanutí:12 °C
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS05, GHS07, GHS09
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Caution: P210 - P273 - P280 - P301 + P310 - P305 + P351 + P338 - P331