ContactUsLinkComponent
Organic reference standard, Phenethyl acetate, analytical standard, Supelco®
Organic reference standard, Phenethyl acetate, analytical standard, Supelco®
Dodavatel:  Merck
Certifikáty
undefined
Organic reference standard, Phenethyl acetate, analytical standard, Supelco®
Dodavatel:  Merck
Specifikace
  • Čí. CAS:
    103-45-7
  • Vzorec:
    CH₃COOCH₂CH₂C₆H₅
  • Popis:
    Phenethyl acetate
  • Bod vzplanutí:
    105 °C
  • Bod varu:
    238…239 °C