Organic reference standard, mono-Butyl phthalate, analytical standard, Supelco®
Catalog # 30751-100MG
Supplier: Merck
CAS Number:
Specifications
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:131-70-4
- Popis:mono-Butyl phthalate
- Vzorec:C₁₂H₁₄O₄
- Popis:mono-Butyl phthalate
- Tvar:Neat