Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, ethyl salicylate, analytical standard, Supelco®
  68291-1ML
 :  Merck
 :  
undefined
Organic reference standard, ethyl salicylate, analytical standard, Supelco®
  68291-1ML
 :  Merck
 :  

 

  • Bal.:
    1 mL
  • Standard:
    Analytical standard
  • Čí. CAS:
    118-61-6
  • Popis:
    Ethyl salicylate, ≥98,0% (GC)
  • Bod tání:
    1 °C (lit.)
  • Vzorec:
    C₉H₁₀O₃
  • Popis:
    Ethyl salicylate
  • Bod varu:
    234 °C (lit.)