Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Diethyl phthalate-3,4,5,6-d4, analytical standard, Supelco®
Organic reference standard, Diethyl phthalate-3,4,5,6-d4, analytical standard, Supelco®
Catalog # 34185-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Diethyl phthalate-3,4,5,6-d4, analytical standard, Supelco®
Catalog # 34185-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Bal.:
    25 mg
  • Standard:
    Analytical standard
  • Čí. CAS:
    93952-12-6
  • Popis:
    Diethyl phthalate-3,4,5,6-d4
  • Bod tání:
    –3 °C (lit.)
  • Vzorec:
    C₁₂H₁₄O₄
  • Popis:
    Diethyl phthalate-3,4,5,6-d4
  • Bod varu:
    298...299 °C (lit.)
  • Tvar:
    Neat

Specifications

About this item