Supelco® Organic Reference Standard, β-Alanine
41144-100MG
: Merck
:
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:107-95-9
- Popis:β-Alanine
- Podmínky uskladnění:2...30 °C
- Podmínky k odeslání:STD
- MDL čí.:MFCD00008200
- M-Clarity quality level:MQ100
- Synonym(s):β-Ala; 3-Aminopropionic acid
- Bod tání:202 °C (dec.) (lit.)
- Vzorec:NH₂CH₂CH₂COOH
- Molekulární hmotnost:89,09
- Bod vzplanutí:Not applicable
- Tvar:Neat
β-Alanine flux plays a cytoprotective role by supporting the osmotic stability of marine organisms, preimplantation mouse embryos and mammalian cells exposed to hypoxic stress.