Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, Aphidicolin, analytical standard, Supelco®
Catalog # 89458-5MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Aphidicolin, analytical standard, Supelco®
Catalog # 89458-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Bal.:
    5 mg
  • Standard:
    Analytical standard
  • Čí. CAS:
    38966-21-1
  • Popis:
    Aphidicolin, ≥95% (HPLC)
  • Bod tání:
    >300 °C (lit.)
  • Vzorec:
    C₂₀H₃₄O₄
  • Skladovací teplota:
    2...8 °C
  • Popis:
    Aphidicolin
  • Tvar:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle contents are inside inserted with fused cone.