Organic reference standard, Amarouciaxanthin A, analytical standard, Supelco®
Catalog # 54758-1MG
Supplier: Merck
CAS Number:
Specifications
- Bal.:1 mg
- Standard:Analytical standard
- Čí. CAS:100667-78-5
- Popis:Amarouciaxanthin A, ≥97,0% (HPLC)
- Vzorec:C₄₀H₅₄O₅
- Skladovací teplota:–20 °C
- Popis:Amarouciaxanthin A