Organic reference standard, Aflatoxin Mix 4 solution, analytical standard, Supelco®
Katalogové číslo 33415-2ML
Dodavatel: Merck
Číslo CAS:
Specifikace
- Bal.:2 mL
- Standard:Analytical standard
- Popis:Aflatoxin Mix 4 solution
- M-Clarity quality level:MQ100
- Popis:Aflatoxin Mix 4 solution
Specifikace
O tomto produktu
Packaging: B1, G1, B2, G2 in acetonitrile (20 ug/ml each).