Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, 6-Acetyl-1,1,2,4,4,7-hexamethyltetralin, analytical standard, Supelco®
Organic reference standard, 6-Acetyl-1,1,2,4,4,7-hexamethyltetralin, analytical standard, Supelco®
Catalog # 54730-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 6-Acetyl-1,1,2,4,4,7-hexamethyltetralin, analytical standard, Supelco®
Catalog # 54730-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Bal.:
    100 mg
  • Standard:
    Analytical standard
  • Čí. CAS:
    21145-77-7
  • Popis:
    6-Acetyl-1,1,2,4,4,7-hexamethyltetralin, ≥97,0% (GC)
  • Vzorec:
    C₁₈H₂₆O
  • Popis:
    6-Acetyl-1,1,2,4,4,7-hexamethyltetralin

Specifications

About this item