ContactUsLinkComponent
Organic reference standard, 3,4-Dihydroxybenzaldehyde, analytical standard, Supelco®
Organic reference standard, 3,4-Dihydroxybenzaldehyde, analytical standard, Supelco®
Catalog # 92588-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 3,4-Dihydroxybenzaldehyde, analytical standard, Supelco®
Catalog # 92588-100MG
Supplier:  Merck
CAS Number:  
Specifications
  • Bal.:
    100 mg
  • Standard:
    Analytical standard
  • Čí. CAS:
    139-85-5
  • Popis:
    3,4-Dihydroxybenzaldehyde, ≥95,0% (HPLC)
  • Bod tání:
    150...155 °C
    150...157 °C (lit.)
  • Vzorec:
    C₇H₆O₃
  • Popis:
    3,4-Dihydroxybenzaldehyde
  • Tvar:
    Neat