0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
Di-2-ANEPEQ (JPW 1114)
Di-2-ANEPEQ (JPW 1114) membrane stain, for histology
Di-2-ANEPEQ (JPW 1114) membrane stain, for histology
Di-2-ANEPEQ (JPW 1114) membrane stain, for histology
Catalog # 61013.
Supplier:
Biotium
CAS Number:
Expand image
Di-2-ANEPEQ (JPW 1114) membrane stain, for histology
Catalog # 61013.
Supplier:
Biotium
CAS Number:
Specifications
Bal.:
5 mg
Sekvence:
2210
Specifications
About this item
Product Details
:
:
549.384 g/mol
Storage Temperature:
Freezer
Formula:
C₂₆H₃₅Br₂N₃
Product Details