Zadání objednávky
Czech Republic
ContactUsLinkComponent
10-Acetyl-3,7-dihydroxyphenoxazine peroxidase substrate
10-Acetyl-3,7-dihydroxyphenoxazine peroxidase substrate
Catalog # 10061.
Supplier:  Biotium
CAS Number:  
undefined
10-Acetyl-3,7-dihydroxyphenoxazine peroxidase substrate
Catalog # 10061.
Supplier:  Biotium
CAS Number:  

Specifications

  • Bal.:
    5 mg
  • Sekvence:
    2210

Specifications

About this item

 :
 :257.24 g/mol
Storage Temperature:Freezer
CAS Number:119171-73-2
Formula:C₁₄H₁₁NO₄
MDL Number:MFCD00467861