Supelco® Organic Reference Standard, F.A.M.E. Mix RM-1
Catalog # O7006-1AMP
Supplier: Merck
CAS Number:
Specifications
- Bal.:1 Ampoul
- Standard:Certified reference material
- Popis:F.A.M.E. mix RM-1
- Podmínky uskladnění:–20 °C
- Popis rizikové třídy:H413
- Podmínky k odeslání:DICE
- M-Clarity quality level:MQ100
- Synonym(s):AOCS Reference Mixtures; AOCS No. 1 (RM-1); Oil Reference Standard, AOCS No. 1; Oil Reference Standard, AOCS
- Pk Info:Ampule of 100 mg (Neat)
Specifications
About this item
Palmitic acid methyl ester (C16:0) 6,0% Stearic acid methyl ester (C18:0) 3,0% Oleic acid methyl ester (C18:1cis-9) 35,0% Linoleic acid methyl ester (C18:2cis-912) 50,0% Linolenic acid methyl ester (C18:3cis-91215) 3,0% Arachidic acid methyl ester (C20:0) 3,0%.
Caution: P273 - P501