Magnetic stirring bar, octagonal, PTFE
Catalog # COWIFT.8360
Supplier: COWIE
Specifications
- Ø:13 mm
- Délka:75 mm
- Bal.:1
Specifications
About this item
Octahedral magnetic stirring bars.
- Octahedral PTFE Stirrer Bar with champhered magnet core
- 100% visual inspection using 4× magnification
Delivery: Supplied with a free of charge certificate of analysis confirming the Job Number, material batch number, date of manufacture and inspection criteria.