Supelco® Organic Reference Standard, EPA 8240B/8260A Matrix Spike Mix
Catalog # 47411-U
Supplier: Merck
CAS Number:
Specifications
- Koncentrace:2500 μg/ml each component in methanol
- Bal.:1
- Standard:Certified reference material
- Popis:EPA 8240B/8260A Matrix spike mix
- Podmínky uskladnění:2…8 °C
- Popis rizikové třídy:H225 - H301 + H311 + H331 - H340 - H350 - H370
- Podmínky k odeslání:STD
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS06, GHS08
Caution: P202 - P210 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311