Chamber slides, C-Slide
Katalogové číslo NANOCS-050
Dodavatel: NANOENTEK
Specifikace
- Doporučený pracovní objem:10 µl
- Popis:C-Slide cell counting chamber slides (100 tests)
- Bal.:50
Specifikace
O tomto produktu
Cell counting chamber slides for use with automated cell counter (Countess and Countess II).
- No need of coverslips
- Eliminates the need of washing and reuse
- Reduced exposures to the infectious samples and contamination
- Guaranteed reproducibility and reliability
Made of sturdy and strong quartz grade optical plastic.