Cleaning swabs, sealed, CONSTIX® SF-9
Katalogové číslo CONTSF-9
Dodavatel: CONTEC
Specifikace
- Popis:Sealed foam swabs, head: 3,6×7,6 mm, handle Ø 2,5 mm
- Délka:66 mm
- Shaft/head:PP/PU
- Bal.:500
Specifikace
O tomto produktu
This sealed foam swab consists of polyurethane foam bonded to a polypropylene handle without the use of adhesives or binders.
- Mini foam head enables good access to small features and difficult to reach surfaces
- Polyurethane foam head provides good particle entrapment and removal
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. Excellent for applying resins and adhesives.
Though compatible with many common solvents, the SF-9 is not recommended for use with ketones such as acetone or MEK.
Balení: Swabs are packed 500 per case.
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904