Supelco® Organic Reference Standard, Methyl Heptadecanoate-D₃₃
Catalog # 00889-10MG
Supplier: Merck
CAS Number:
Specifications
- Bal.:10 mg
- Standard:Analytical standard
- Čí. CAS:1219804-81-5
- Popis:Methyl heptadecanoate-d₃₃
- Podmínky uskladnění:RT
- Podmínky k odeslání:STD
- MDL čí.:MFCD03428281
- M-Clarity quality level:MQ200
- Synonym(s):Heptadecanoic acid-d₃₃ methyl ester
- Vzorec:C₁₈D₃₃H₃O₂
- Molekulární hmotnost:317,68
- Bod vzplanutí:Not applicable
Specifications
About this item
Packaging: Bottomless glass bottle contents are inside inserted fused cone.