Organic reference standard, Ethylhexyl triazone, analytical standard, Supelco®
Catalog # 93465-100MG
Supplier: Merck
CAS Number:
Specifications
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:88122-99-0
- Popis:Ethylhexyl triazone
- Bod tání:128...132 °C
- Vzorec:C₄₈H₆₆N₆O₆
- Popis:Ethylhexyl triazone
Specifications
About this item
Ethylhexyl triazone is a ultraviolet-B filter, which is used as a sunscreen agent in skin care products.
Packaging: Bottomless glass bottles are inserted with fused cone.