Organic reference standard, 2-Ethylhexyl 4-methoxycinnamate, analytical standard, Supelco®
Catalog # 55529-100MG
Supplier: Merck
CAS Number:
Specifications
- Bal.:100 mg
- Standard:Analytical standard
- Čí. CAS:5466-77-3
- Popis:2-Ethylhexyl 4-methoxycinnamate
- Vzorec:C₁₈H₂₆O₃
- Popis:2-Ethylhexyl 4-methoxycinnamate
Specifications
About this item
2-Ethylhexyl 4-methoxycinnamate is a commonly used chemical UV filter, which can generally find applications as a sunscreen ingredient.