Stopcocks, three-way
Stopcock, three-way
Katalogové číslo SIALCAD6001-1EA
Dodavatel: Merck
Přejít na:
Product Details & Documents
The spring clip design holds the key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
- Made from nickel plated brass or stainless steel
- Tested to 45 psi water pressure
- High pressure stopcocks are rated to 290 psi
Female luer lock to male luer, right side, female Luer lock, compression-nut.
Specifikace
- Popis:Stopcock, three-way