Vacuum pump, EZ-Stream
EZ-Stream vacuum pump for liquid transfer, W×D×H: 196×168×222 mm, 1,9 kg
Katalogové číslo MILYEZSTREAM1
New Product
Dodavatel: MILLIPORE
Přejít na:
Product Details & Documents
This laboratory filtration pump is ideal to transfer filtered liquids directly to waste.
- Flow rate: 3,8 to 4,0 L/min
Used to transfer beer, dairy, bottled water, food and beverage, pharmaceutical products analysis, soft drink, cider, sport drink and wine.
Delivery information: Includes pump, multilingual user guide and power supply.
Specifikace
- Popis:EZ-Stream vacuum pump for liquid transfer, W×D×H: 196×168×222 mm, 1,9 kg