Gas burette, Bunte, Class A
WITG2.970.000
- Kapacita:100 ml
- Dělení:0,2 ml
- Bal.:1
Gas burette made of borosilicate glass, accuracy Class A.
- With resistant DIFFICO graduation
- Main point ring graduation
ST-stopcock with double bore and screw-thread retaining nut.