Laboratory bottles, narrow neck, without screw cap
Catalog # KAVA632414375100
Supplier: Kavalier
Specifications
- Ø vněj.:56 mm
- Kapacita:100 ml
- Výška:105 mm
- Sekvence:01
- Bal.:10
Specifications
About this item
Borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Certifications: In compliance with DIN ISO 4796 - 1 standards