Colorimetric rapid tests, manganese, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Certificates
About this item
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
- Portable
- Environmentally friendly
- Convenient handling
- Application-oriented measuring ranges
Natural waters contain manganese in the divalent, soluble form as well as in colloidal tri- and tetravalent states. Valencies are interchanged by oxidation-reduction reactions taking place in the water. The test procedure determines all oxidation states of manganese.
Hazardous material: Yes
Specifications
- Model:MANA920155
- Plošina:Colorimetric test kits
- Skladovací teplota:15...25 °C
- Životnost police (měsíce):18
- Hmotnost:193,57 g (Including packaging)