Orders LinkContactUsLinkComponent
Erlenmeyer flasks, polymethylpentene
Erlenmeyer flasks, polymethylpentene
02, 250 ml
Katalogové číslo HACH2089846
Dodavatel:   Hach
Erlenmeyer flasks, polymethylpentene
Katalogové číslo HACH2089846
Dodavatel:  Hach
Erlenmeyer flasks, polymethylpentene
Erlenmeyer flasks, polymethylpentene
02, 250 ml
Přejít na:
Price
1 120,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Ideal for titrations, preparation and storage of media.
  • Chemical resistance Strength
  • Autoclavable
Polymethylpentene flask with polypropylene screwcap features clarity (Do not engage threads during cycle).
Specifikace
  • Kapacita:
    250 ml
  • Sekvence:
    02