Flexible coupling for stirrers
Catalog # WITG1.445.500
Supplier: witeg Labortechnik
Specifications
- Popis:Flexible coupling for stirrers
- Environmentally Preferable:
- Bal.:1
- Shaft Ø:8 mm
Specifications
About this item
This flexible coupling fix between motor and stirrer shaft.
- Made of PTFE/polyamid