0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Standardy
Standardy pro environmentální analýzu
PAHs by HPLC, (CRM), Supelco®
PAHs by HPLC, (CRM), Supelco®
Catalog # SQC017-40G
Supplier:
Merck
CAS Number:
Expand image
PAHs by HPLC, (CRM), Supelco®
Catalog # SQC017-40G
Supplier:
Merck
CAS Number:
Specifications
Bal.:
40 g
Popis:
PAHs by HPLC
Typ:
Certified reference material
Specifications
About this item
Product Details
Product Details