0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
Standardy
Standardy pro environmentální analýzu
Low level - complex nutrients, (CRM), Supelco®
Low level - complex nutrients, (CRM), Supelco®
Low level - complex nutrients, (CRM), Supelco®
Catalog # RTC000014-2ML
Supplier:
Merck
CAS Number:
Expand image
Low level - complex nutrients, (CRM), Supelco®
Catalog # RTC000014-2ML
Supplier:
Merck
CAS Number:
Specifications
Bal.:
2 mL
Popis:
Low Level - Complex Nutrients
Specifications
About this item
Product Details
Product Details