Supelco® Organic Reference Standard, Loam Matrix Blank
Catalog # MB010-50G
Supplier: Merck
CAS Number:
Specifications
- Bal.:50 g
- Standard:Certified reference material
- Popis:Loam matrix blank
- Podmínky uskladnění:2...30 °C
- Podmínky k odeslání:STD
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS08