ContactUsLinkComponent
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥95% (by HPLC) fluorescent indicator, for chloride determination
Irritant
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥95% (by HPLC) fluorescent indicator, for chloride determination
Catalog # 52011.
Supplier:  Biotium
CAS Number:  
undefined
Irritant
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥95% (by HPLC) fluorescent indicator, for chloride determination
Catalog # 52011.
Supplier:  Biotium
CAS Number:  
Specifications
  • Bal.:
    100 mg
  • Sekvence:
    2440
Specifications
About this item
 :
 :326.19 g/mol
 :GHS07
CAS Number:162558-52-3
Formula:C₁₄H₁₆NO₃Br
MDL Number:MFCD00467854