Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standardy
Standardy organické
Pharmaceutical secondary standard, ritonavir, (CRM), Supelco®
Pharmaceutical secondary standard, ritonavir, (CRM), Supelco®
Pharmaceutical secondary standard, ritonavir, (CRM), Supelco®
Catalog # PHR1734-1G
Supplier:
Merck
CAS Number:
Expand image
Pharmaceutical secondary standard, ritonavir, (CRM), Supelco®
Catalog # PHR1734-1G
Supplier:
Merck
CAS Number:
Specifications
Bal.:
1 g
Čí. CAS:
155213-67-5
Popis:
Ritonavir
Popis:
Ritonavir
Specifications
About this item
Product Details
Product Details