Pharmaceutical secondary standard, octocrylene, (CRM), Supelco®
: Merck
- Čí. CAS:6197-30-4
- Popis:Octocrylene
Frequently Bought Together
Tymol Ph. Eur.
: VWR Chemicals
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Aceton ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Ubrousky čisticí
: VWR Collection
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Volumetric flasks, class B, Azlon®
: SciLabware
Benzofenon 99+%, pure
: Thermo Fisher Scientific