Pharmaceutical secondary standard, octisalate, (CRM), Supelco®
Supplier: Merck
Certificates
Certificates
About this item
Specifications
- Čí. CAS:118-60-5
- Popis:Octisalate
Specifications
Frequently Bought Together
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Supplier: VWR Chemicals
VWR®, AVS TITRINORM pH pufrovací roztoky v sáčcích, 20 °C
Supplier: VWR Chemicals
Accessories for pH/°C meter, handheld
Supplier: Testo
VWR® ARISTAR®, Titanium Standard Solutions for ICP-MS
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Pirimicarb Analytical standard, reference material, Fluka™
Supplier: Honeywell Chemicals
[EN]PARACETAMOL IMPURITY J 1 * 15 mg
Supplier: EDQM
TRIFLUMIZOLE-AMINO 1 * 10 mg
Supplier: EHRENSTORFER
FENOXAPROP-P (R-ENANTIOMER) CERTIFIED 1 * 0,1 g
Supplier: EHRENSTORFER
Tlumivé roztoky v sáčcích, perpHect® sáčky, Orion™
Supplier: Thermo Orion
VWR®, High temperature viscosity standards
Supplier: VWR Chemicals
[EN]STRAY LIGHT STANDARD, FOR INSTRUMENT 1 * 1 SET
Supplier: Hach
Plastic Additive, Reference Standards
Supplier: USP
[EN]R-LEVETIRACETAM HYDROXYBUTYRAMIDE AN 1 * 10 mg
Supplier: Merck