0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
Standardy
Standardy organické
Pharmaceutical secondary standard, maltitol, (CRM), Supelco®
Pharmaceutical secondary standard, maltitol, (CRM), Supelco®
Pharmaceutical secondary standard, maltitol, (CRM), Supelco®
Catalog # PHR1248-500MG
Supplier:
Merck
CAS Number:
Expand image
Pharmaceutical secondary standard, maltitol, (CRM), Supelco®
Catalog # PHR1248-500MG
Supplier:
Merck
CAS Number:
Specifications
Bal.:
500 mg
Čí. CAS:
585-88-6
Popis:
Maltitol
Popis:
Maltitol
Specifications
About this item
Product Details
Product Details