Supelco® Organic Reference Standard, Clarithromycin Impurity Q
Catalog # PHR1968-20MG
Supplier: Merck
CAS Number:
Specifications
- Bal.:20 mg
- Standard:Pharmaceutical Secondary Standard; Certified Reference Material
- Čí. CAS:118074-07-0
- Popis:Clarithromycin impurity Q
- Podmínky uskladnění:–10...–25 °C
- Podmínky k odeslání:DICE
- MDL čí.:MFCD24386630
- M-Clarity quality level:MQ100
- Synonym(s):Clarithromycin N-oxide; 6-O-Methylerythromycin N-oxide
- Molekulární hmotnost:763,95