Supelco® TraceCERT® Organic Reference Standard, Bis(2-ethylhexyl) phthalate
67261-100MG
: Merck
:
- Bal.:100 mg
- Standard:Certified reference material, TraceCERT®
- Čí. CAS:117-81-7
- Popis:Bis(2-ethylhexyl) phthalate
- Podmínky uskladnění:15...25 °C
- Popis rizikové třídy:H360FD
- Podmínky k odeslání:STD
- MDL čí.:MFCD00009493
- M-Clarity quality level:MQ300
- Synonym(s):DEHP; Phthalic acid bis(2-ethylhexyl ester); ‘Dioctyl’ phthalate
- Molekulární hmotnost:390,56
- Hustota:0,985 g/ml at 20 °C (lit.)
- Bod vzplanutí:207,0 °C - Closed cup
- Bod varu:386 °C (lit.)
- GHS signal: GHS08
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P201 - P202 - P280 - P308 + P313 - P405 - P501