Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standardy
Standardy organické
Organic reference standard, neat, (+)-Carvone, (CRM)
Organic reference standard, neat, (+)-Carvone, (CRM)
Organic reference standard, neat, (+)-Carvone, (CRM)
Catalog # 79245-100MG
Supplier:
Merck
CAS Number:
Expand image
Organic reference standard, neat, (+)-Carvone, (CRM)
Catalog # 79245-100MG
Supplier:
Merck
CAS Number:
Specifications
Bal.:
100 mg
Čí. CAS:
2244-16-8
Popis:
(+)-Carvone
Popis:
(+)-Carvone
Specifications
About this item
Product Details
Product Details