Supelco® Drug and metabolite standards, Tapentadol hydrochloride solution, (CRM)
T-058-1ML
: Merck
:
- Koncentrace:1,0 mg/ml in methanol (as free base)
- Bal.:1 mL
- Standard:Certified reference material
- Čí. CAS:175591-09-0
- Popis:Tapentadol hydrochloride solution
- Popis:Tapentadol hydrochloride solution
- Pk Info:ampoule of 1 ml