Order Entry
Czech Republic
ContactUsLinkComponent
 
Supelco® Drug and metabolite standards, N-desmethyltapentadol solution, (CRM)
Catalog # D-052-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Drug and metabolite standards, N-desmethyltapentadol solution, (CRM)
Catalog # D-052-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Koncentrace:
    1,0 mg/ml in methanol
  • Bal.:
    1 mL
  • Standard:
    Certified reference material
  • Čí. CAS:
    1246819-18-0
  • Popis:
    N-Desmethyltapentadol solution
  • Popis:
    N-Desmethyltapentadol solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item