Supelco® Drug and metabolite standards, naloxone-3ß-D-glucuronide solution, (CRM)
N-099-1ML
: Merck
:
- Koncentrace:1,0 mg/ml in methanol: water (9:1)
- Bal.:1 mL
- Standard:Certified reference material
- Popis:Naloxone-3ß-D-glucuronide solution
- Pk Info:ampoule of 1 ml