Supelco® Drug and metabolite standards, naloxone solution, (CRM)
Catalog # N-004-1ML
Supplier: Merck
CAS Number:
Specifications
- Koncentrace:1,0 mg/ml in methanol
- Bal.:1 mL
- Standard:Certified reference material
- Čí. CAS:465-65-6
- Popis:Naloxone solution
- Popis:Naloxone solution
- Pk Info:ampoule of 1 ml